Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-1,2-Cyclohexanedicarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A240468-100G A240468-10G A240468-500G
76815-396EA 18 USD InStock
76815-396 77672-142 77672-144
cis-1,2-Cyclohexanedicarboxylic acid 98%
cis-1,2-Cyclohexanedicarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₂O₄
MDL Number: MFCD00064949
CAS Number: 610-09-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty