Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-Pyrazol-5-yl)aniline 98%

Supplier: AMBEED, INC
Warning

A247205-100MG A247205-1G A247205-250MG
76815-550EA 23 USD InStock
76815-550 76815-552 76815-554
2-(1H-Pyrazol-5-yl)aniline 98%
2-(1H-Pyrazol-5-yl)aniline
Precaución: for Research Use Only


Formula: C₉H₉N₃
MDL Number: MFCD09028476
CAS Number: 111562-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty