Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylmorpholine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A253931-100MG A253931-1G A253931-250MG A253931-5G
76815-750EA 96 USD InStock
76815-750 76815-752 76815-754 76815-756
2-Phenylmorpholine hydrochloride 95%
2-Phenylmorpholine hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO·ClH
MDL Number: MFCD03869024
CAS Number: 23972-42-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty