Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-tert-Butyl (3-oxocyclopentyl)carbamate 95%

Supplier: AMBEED, INC
Warning

A255975-250MG A255975-100MG
76815-844EA 39 USD InStock
76815-844 77672-284
(S)-tert-Butyl (3-oxocyclopentyl)carbamate 95%
(S)-tert-Butyl (3-oxocyclopentyl)carbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
MDL Number: MFCD18089809
CAS Number: 167298-40-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty