Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridin-2-ol 95%

Supplier: AMBEED, INC
Warning

A258069-10G A258069-1G A258069-250MG A258069-5G
76815-946EA 161 USD InStock
76815-946 76815-948 76815-950 77672-302
Imidazo[1,2-a]pyridin-2-ol 95%
Imidazo[1,2-a]pyridin-2-ol
Precaución: for Research Use Only


Formula: C₇H₆N₂O
MW: 134.135 g/mol
MDL Number: MFCD09994219
CAS Number: 112566-20-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty