Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-cyanobenzoate 97%

Supplier: AMBEED, INC
Warning

A259523-100G A259523-10G A259523-1G A259523-25G A259523-5G
76815-974EA 168 USD InStock
76815-974 76815-976 76815-978 76815-980 76815-982
Ethyl-4-cyanobenzoate 97%
Ethyl-4-cyanobenzoate
Precaución: for Research Use Only


Formula: C₁₀H₉NO₂
MDL Number: MFCD00001824
CAS Number: 7153-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty