Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R)-2-Methylpyrrolidine-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A261311-1G A261311-250MG A261311-5G A261311-10G A261311-25G
76816-012EA 294 USD InStock
76816-012 76816-014 76816-016 77672-332 77672-334
(2R)-2-Methylpyrrolidine-2-carboxylic acid 97%
(2R)-2-Methylpyrrolidine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₁₁NO₂
MDL Number: MFCD08274534
CAS Number: 63399-77-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty