Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

JNJ-7706621 ≥98%

Supplier: AMBEED, INC
Warning

A262123-10MG A262123-1MG A262123-2MG A262123-50MG A262123-5MG
76816-024EA 69 USD InStock
76816-024 76816-026 76816-028 76816-030 76816-032
JNJ-7706621 ≥98%
JNJ-7706621
Precaución: for Research Use Only


Formula: C₁₅H₁₂F₂N₆O₃S
MDL Number: MFCD11100270
CAS Number: 443797-96-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty