Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5,6-Triaminopyrimidin-4(3H)-one sulfate 95%

Supplier: AMBEED, INC
Warning

A269818-1000G A269818-100G A269818-500G A269818-25G A269818-5G
76816-166EA 129 USD InStock
76816-166 76816-168 76816-170 77092-650 77612-578
2,5,6-Triaminopyrimidin-4(3H)-one sulfate 95%
2,5,6-Triaminopyrimidin-4(3H)-one sulfate
Precaución: for Research Use Only


Formula: C₄H₉N₅O₅S
MW: 239.20 g/mol
MDL Number: MFCD23099603
CAS Number: 35011-47-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty