Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methyl-2,2'-bipyridine 98%

Supplier: AMBEED, INC
Warning

A272877-100MG A272877-1G A272877-250MG
76816-238EA 21 USD InStock
76816-238 76816-240 76816-242
6-Methyl-2,2'-bipyridine 98%
6-Methyl-2,2'-bipyridine
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂
MDL Number: MFCD02093688
CAS Number: 56100-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty