Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-DL-alaninol 97%

Supplier: AMBEED, INC
Warning

A273445-100G A273445-10G A273445-1G A273445-250MG A273445-25G A273445-5G
76816-254EA 79 USD InStock
76816-254 76816-256 76816-258 76816-260 76816-262 76816-264
N-Boc-DL-alaninol 97%
N-Boc-DL-alaninol
Precaución: for Research Use Only


Formula: C₈H₁₇NO₃
MDL Number: MFCD04973335
CAS Number: 147252-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty