Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Terazosin hydrochloride 99%

Supplier: AMBEED, INC
Warning

A283609-100G A283609-1G
76816-404EA 356 USD InStock
76816-404 76816-406 76816-408 76816-410
Terazosin hydrochloride 99%
Terazosin hydrochloride
Precaución: for Research Use Only


Formula: C₁₉H₂₅N₅O₄·HCl
MW: 423.90 g/mol
MDL Number: MFCD00467965
CAS Number: 63074-08-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty