Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(α)-Boc-DL-2,3-diaminopropionic acid 95%

Supplier: AMBEED, INC
Warning

A295186-10G A295186-1G A295186-250MG
76816-642EA 25 USD InStock
76816-642 76816-644 76816-646
N-(α)-Boc-DL-2,3-diaminopropionic acid 95%
N-(alpha)-Boc-DL-2,3-diaminopropionic acid
Precaución: for Research Use Only


Formula: C₈H₁₆N₂O₄
MW: 204.23 g/mol
MDL Number: MFCD00736752
CAS Number: 159002-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty