Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-methylphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A298802-100MG A298802-1G A298802-250MG A298802-5G
76816-714EA 8 USD InStock
76816-714 76816-716 76816-718 76816-720
3-Chloro-5-methylphenylboronic acid 97%
3-Chloro-5-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BClO₂
MDL Number: MFCD09027241
CAS Number: 913836-14-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty