Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloronicotinic acid 97%

Supplier: AMBEED, INC
Warning

A304452-1G A304452-25G A304452-5G
76816-812EA 14 USD InStock
76816-812 76816-814 76816-816
2,5-Dichloronicotinic acid 97%
2,5-Dichloronicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃Cl₂NO₂
MW: 192
MDL Number: MFCD01861979
CAS Number: 59782-85-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty