Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,6-Dichloro-3-methoxyphenyl)boronic acid 95%

Supplier: AMBEED, INC
Warning

A305054-100MG A305054-1G A305054-250MG A305054-5G
76816-820EA 23 USD InStock
76816-820 76816-822 76816-824 76816-826
(2,6-Dichloro-3-methoxyphenyl)boronic acid 95%
(2,6-Dichloro-3-methoxyphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₇H₇BCl₂O₃
MW: 220.84 g/mol
MDL Number: MFCD11865199
CAS Number: 851756-57-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty