Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-cyanobenzeneboronic acid 94%

Supplier: AMBEED, INC
Warning

A315055-1G A315055-250MG A315055-5G
76816-930EA 27 USD InStock
76816-930 76816-932 76816-934
5-Bromo-2-cyanobenzeneboronic acid 94%
5-Bromo-2-cyanobenzeneboronic acid
Precaución: for Research Use Only


Formula: C₇H₅BBrNO₂
MW: 225.836 g/mol
MDL Number: MFCD10696674
CAS Number: 1032231-30-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty