Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 95%

Supplier: AMBEED, INC
Danger

A327311-250MG A327311-5G A327311-1G
76817-124EA 14 USD InStock
76817-124 76817-126 77672-682
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 95%
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid
Precaución: for Research Use Only


Formula: C₄H₅F₃O₃
MW: 158.08 g/mol
MDL Number: MFCD00190646
CAS Number: 374-35-6
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty