Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methoxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A334106-1G A334106-250MG A334106-5G
76817-198EA 13 USD InStock
76817-198 76817-200 76817-202
2-Chloro-5-methoxybenzaldehyde 97%
2-Chloro-5-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
MDL Number: MFCD11845925
CAS Number: 13719-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty