Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Spiro[isochroman-1,4'-piperidine] hydrochloride 95%

Supplier: AMBEED, INC
Warning

A343797-100MG A343797-250MG
76817-262EA 40 USD InStock
76817-262 76817-264
Spiro[isochroman-1,4'-piperidine] hydrochloride 95%
Spiro[isochroman-1,4'-piperidine] hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₁₈ClNO
MDL Number: MFCD20134157
CAS Number: 173943-98-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty