Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bisbenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A347350-100G A347350-25G A347350-500G A347350-5G
76817-278EA 51 USD InStock
76817-278 76817-280 76817-282 76817-284
1,3-Bisbenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid 97%
1,3-Bisbenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₁₉H₁₈N₂O₅
MDL Number: MFCD06798134
CAS Number: 59564-78-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty