Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromoquinoline hydrochloride 97%

Supplier: AMBEED, INC
Warning

A358518-100G A358518-25G A358518-5G
76817-400EA 51 USD InStock
76817-400 76817-402 76817-404
6-Bromoquinoline hydrochloride 97%
6-Bromoquinoline hydrochloride
Precaución: for Research Use Only


Formula: C₉H₇BrClN
MDL Number: MFCD12756475
CAS Number: 55377-26-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty