Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-ethynylpyridine 95%

Supplier: AMBEED, INC
Warning

A373239-100MG A373239-250MG
76817-534EA 34 USD InStock
76817-534 76817-536
2-Bromo-6-ethynylpyridine 95%
2-Bromo-6-ethynylpyridine
Precaución: for Research Use Only


Formula: C₇H₄BrN
MDL Number: MFCD13189718
CAS Number: 445468-63-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty