Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',4'-Dichlorovalerophenone 98+%

Supplier: AMBEED, INC
Warning

A374081-100G A374081-25G A374081-5G
76817-568EA 180 USD InStock
76817-568 76817-570 76817-572
2',4'-Dichlorovalerophenone 98+%
2',4'-Dichlorovalerophenone
Precaución: for Research Use Only


Formula: C₁₁H₁₂Cl₂O
MDL Number: MFCD09028061
CAS Number: 61023-66-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty