Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carboxy-2-methoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A380460-100MG A380460-1G A380460-250MG A380460-5G
76817-652EA 31 USD InStock
76817-652 76817-654 76817-656 76817-658
3-Carboxy-2-methoxyphenylboronic acid 98%
3-Carboxy-2-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₉BO₅
MDL Number: MFCD09027237
CAS Number: 913836-10-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty