Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)-4-methylpyridine hydrochloride 96%

Supplier: AMBEED, INC
Danger

A384003-100MG A384003-1G A384003-250MG
76817-686EA 23 USD InStock
76817-686 76817-688 76817-690
2-(Chloromethyl)-4-methylpyridine hydrochloride 96%
2-(Chloromethyl)-4-methylpyridine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₉Cl₂N
MW: 178.0600 g/mol
MDL Number: MFCD18384640
CAS Number: 71670-71-8
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty