Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-chlorobenzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A394424-100G A394424-10G A394424-1G A394424-25G A394424-5G
76817-810EA 110 USD InStock
76817-810 76817-812 76817-814 76817-816 76817-818
3-Bromo-4-chlorobenzyl alcohol 97%
3-Bromo-4-chlorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₆BrClO
MW: 221.479 g/mol
MDL Number: MFCD08059085
CAS Number: 329218-12-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty