Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclopropyl-1H-pyrazol-5-amine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A401743-10G A401743-5G
76817-882EA 280 USD InStock
76817-882 76817-884
3-Cyclopropyl-1H-pyrazol-5-amine hydrochloride 95%
3-Cyclopropyl-1H-pyrazol-5-amine hydrochloride
Precaución: for Research Use Only


Formula: C6H10ClN3
MDL Number: MFCD08702764
CAS Number: 1031791-16-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty