Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1,10-Phenanthroline)(trifluoromethyl)copper(I) 90%

Supplier: AMBEED, INC
Warning

A404160-100MG A404160-10G A404160-1G A404160-250MG A404160-25G A404160-5G
76817-912EA 14 USD InStock
76817-912 76817-914 76817-916 76817-918 76817-920 76817-922
(1,10-Phenanthroline)(trifluoromethyl)copper(I) 90%
(1,10-Phenanthroline)(trifluoromethyl)copper(I)
Precaución: for Research Use Only


Formula: C₁₃H₈CuF₃N₂
MDL Number: MFCD19690901
CAS Number: 1300746-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty