Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-fluorobenzylamine 97%

Supplier: AMBEED, INC
Danger

A407173-10G A407173-1G A407173-250MG A407173-5G
76817-938EA 44 USD InStock
76817-938 76817-940 76817-942 77672-984
4-Chloro-3-fluorobenzylamine 97%
4-Chloro-3-fluorobenzylamine
Precaución: for Research Use Only


Formula: C₇H₇ClFN
MW: 159.59 g/mol
MDL Number: MFCD04116008
CAS Number: 72235-58-6
UN: 2735
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty