Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1-Boc-5-methoxy-1,2-benzenediamine 96%

Supplier: AMBEED, INC
Warning

A409989-100MG A409989-250MG
76817-970EA 35 USD InStock
76817-970 76817-972
N1-Boc-5-methoxy-1,2-benzenediamine 96%
N1-Boc-5-methoxy-1,2-benzenediamine
Precaución: for Research Use Only


Formula: C₁₂H₁₈N₂O₃
MDL Number: MFCD09701255
CAS Number: 362670-09-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty