Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl imidazo[1,2-a]pyridine-8-carboxylate 98%

Supplier: AMBEED, INC
Warning

A415626-1G A415626-5G
76818-010EA 18 USD InStock
76818-010 76818-012
Methyl imidazo[1,2-a]pyridine-8-carboxylate 98%
Methyl imidazo[1,2-a]pyridine-8-carboxylate
Precaución: for Research Use Only


Formula: C₉H₈N₂O₂
MDL Number: MFCD09842621
CAS Number: 133427-07-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty