Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate 98%

Supplier: AMBEED, INC
Warning

A430458-100MG A430458-1G A430458-250MG A430458-5G
76818-116EA 5 USD InStock
76818-116 76818-118 76818-120 76818-122
Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate 98%
Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate
Precaución: for Research Use Only


Formula: C₈H₈O₆S
MW: 232.21 g/mol
MDL Number: MFCD01860087
CAS Number: 58416-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty