Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-3-oxobutanoate 95%

Supplier: AMBEED, INC
Danger

A430766-100MG A430766-250MG
76818-128EA 176 USD InStock
76818-128 76818-130
Methyl 2-bromo-3-oxobutanoate 95%
Methyl 2-bromo-3-oxobutanoate
Precaución: for Research Use Only


Formula: C₅H₇BrO₃
MDL Number: MFCD20621408
CAS Number: 3600-18-8
UN: 1760
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty