Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromo-5-formyl-2-methoxyphenyl)boronic acid 96%

Supplier: AMBEED, INC
Danger

A435608-1G A435608-250MG A435608-5G
76818-188EA 66 USD InStock
76818-188 76818-190 76818-192
(3-Bromo-5-formyl-2-methoxyphenyl)boronic acid 96%
(3-Bromo-5-formyl-2-methoxyphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₈H₈BBrO₄
MW: 258.86 g/mol
MDL Number: MFCD09265141
CAS Number: 1072951-80-4
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty