Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-aminocrotonate 98%

Supplier: AMBEED, INC
Warning

A437352-1000G A437352-100G A437352-10G A437352-25G A437352-500G A437352-5G
76818-214EA 83 USD InStock
76818-214 76818-216 76818-218 76818-220 76818-222 76818-224
Ethyl-3-aminocrotonate 98%
Ethyl-3-aminocrotonate
Precaución: for Research Use Only


Formula: C₆H₁₁NO₂
MDL Number: MFCD00008073
CAS Number: 7318-00-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty