Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',7'-Dichlorofluorescin diacetate 97%

Supplier: AMBEED, INC
Warning

A439906-1MG A439906-250MG A439906-50MG
76818-238EA 7 USD InStock
76818-238 77613-350 77673-080
2',7'-Dichlorofluorescin diacetate 97%
2',7'-Dichlorofluorescin diacetate
Precaución: for Research Use Only


Formula: C₂₄H₁₆Cl₂O₇
MDL Number: MFCD00128955
CAS Number: 4091-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty