Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-chloro-5-iodobenzoate 98%

Supplier: AMBEED, INC
Warning

A460860-250MG A460860-25G A460860-5G
76818-412EA 16 USD InStock
76818-412 77673-176 77673-178
Methyl-3-chloro-5-iodobenzoate 98%
Methyl 3-chloro-5-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₆ClIO₂
MDL Number: MFCD00673018
CAS Number: 289039-85-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty