Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Chloroacetophenone 95%

Supplier: AMBEED, INC
Warning

A463389-1000G A463389-100G A463389-25G A463389-500G
76818-428EA 147 USD InStock
76818-428 76818-430 76818-432 76818-434 76818-436
2'-Chloroacetophenone 95%
2'-Chloroacetophenone
Precaución: for Research Use Only


Formula: C₈H₇ClO
MDL Number: MFCD00000560
CAS Number: 2142-68-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty