Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(trifluoromethyl)benzyl bromide 95%

Supplier: AMBEED, INC
Danger

A470743-1G A470743-250MG A470743-5G
76818-518EA 64 USD InStock
76818-518 76818-520 76818-522
4-Chloro-2-(trifluoromethyl)benzyl bromide 95%
4-Chloro-2-(trifluoromethyl)benzyl bromide
Precaución: for Research Use Only


Formula: C₈H₅BrClF₃
MDL Number: MFCD04115840
CAS Number: 886496-75-9
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty