Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Iodo-1H-indole 95%

Supplier: AMBEED, INC
Danger

A498285-1G A498285-250MG
76818-722EA 263 USD InStock
76818-722 77673-302
7-Iodo-1H-indole 95%
7-Iodo-1H-indole
Precaución: for Research Use Only


Formula: C₈H₆IN
MDL Number: MFCD10000889
CAS Number: 89976-15-8
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty