Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-3-formylphenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A506230-1G A506230-250MG A506230-5G
76818-766EA 30 USD InStock
76818-766 76818-768 76818-770
2,4-Difluoro-3-formylphenylboronic acid 95%
2,4-Difluoro-3-formylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₅BF₂O₃
MW: 185.9200 g/mol
MDL Number: MFCD04974116
CAS Number: 870718-06-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty