Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-isopropylbenzoic acid 95%

Supplier: AMBEED, INC
Warning

A518942-100MG A518942-1G A518942-250MG A518942-5G
76818-856EA 36 USD InStock
76818-856 76818-858 76818-860 76818-862
2-Amino-5-isopropylbenzoic acid 95%
2-Amino-5-isopropylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂
MW: 179.21 g/mol
MDL Number: MFCD02358894
CAS Number: 68701-22-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty