Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-amino-3-mercaptopropanoate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A530654-100G A530654-100MG A530654-1G A530654-250MG A530654-25G A530654-5G
76818-916EA 277 USD InStock
76818-916 76818-918 76818-920 76818-922 76818-924 76818-926
Ethyl 2-amino-3-mercaptopropanoate hydrochloride 97%
Ethyl 2-amino-3-mercaptopropanoate hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClNO₂S
MDL Number: MFCD00173796
CAS Number: 93964-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty