Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-formylbenzeneboronic acid 95%

Supplier: AMBEED, INC
Danger

A545037-100MG A545037-1G A545037-250MG A545037-5G
76819-000EA 10 USD InStock
76819-000 76819-002 76819-004 76819-006
3-Fluoro-5-formylbenzeneboronic acid 95%
3-Fluoro-5-formylbenzeneboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BFO₃
MW: 167.93 g/mol
MDL Number: MFCD11856017
CAS Number: 328956-60-1
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty