Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-3-(Boc-amino)cyclobutanecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A554831-100MG A554831-1G A554831-250MG A554831-5G
76819-080EA 17 USD InStock
76819-080 76819-082 76819-084 76819-086
trans-3-(Boc-amino)cyclobutanecarboxylic acid 98%
trans-3-(Boc-amino)cyclobutanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
MDL Number: MFCD06657680
CAS Number: 939400-34-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty