Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxy-N-methyl-2-oxopropanamide 95+%

Supplier: AMBEED, INC
Warning

A578598-100MG A578598-1G A578598-250MG
76819-250EA 41 USD InStock
76819-250 76819-252 76819-254
N-Methoxy-N-methyl-2-oxopropanamide 95+%
N-Methoxy-N-methyl-2-oxopropanamide
Precaución: for Research Use Only


Formula: C₅H₉NO₃
MW: 131.13 g/mol
MDL Number: MFCD09029567
CAS Number: 914220-85-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty