Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-β-Citronellol 95%

Supplier: AMBEED, INC
Warning

A585782-100G A585782-10G A585782-1G A585782-25G A585782-500G A585782-5G
76819-280EA 104 USD InStock
76819-280 76819-282 76819-284 76819-286 76819-288 76819-290
(S)-(-)-β-Citronellol 95%
(S)-(-)-β-Citronellol
Precaución: for Research Use Only


Formula: C₁₀H₂₀O
MW: 156.27 g/mol
MDL Number: MFCD00063214
CAS Number: 7540-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty