Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dimethoxy-2,5-dimethylbenzene 95%

Supplier: AMBEED, INC
Warning

A588932-100MG A588932-1G A588932-250MG A588932-5G
76819-314EA 62 USD InStock
76819-314 76819-316 76819-318 76819-320
1,3-Dimethoxy-2,5-dimethylbenzene 95%
1,3-Dimethoxy-2,5-dimethylbenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₄O₂
MDL Number: MFCD09835328
CAS Number: 21390-25-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty