Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylcyclohexanone 97%

Supplier: AMBEED, INC
Danger

A596950-25G A596950-5G A596950-10G
76819-364EA 14 USD InStock
76819-364 76819-366 77673-604
4-Methylcyclohexanone 97%
4-Methylcyclohexanone
Precaución: for Research Use Only


Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 170
Melting Pt: -41
Flash Pt: 48
MDL Number: MFCD00001643
CAS Number: 589-92-4
UN: 1224
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty